Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007589.3 | NP_001007590.2 | 4 | 984 | ||
GenBank mRNA | AK005836.1 | - | 4 | 666 | ||
BC082305.1 | - | 4 | 3344 |