Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002917.1 | NP_002908.1 | 7, 8 | 929 | ||
XM_011523587.2 | XP_011521889.1 | 6, 7 | 623 | |||
GenBank mRNA | AF038196.1 | - | 4, 5 | 503 | ||
AK094016.1 | - | 4, 5 | 1144 | |||
AK293568.1 | - | 8, 9 | 875 | |||
AK302068.1 | - | 7, 8 | 738 | |||
BC014495.1 | - | 3, 4 | 377 | |||
BC069034.1 | - | 3, 4 | 856 | |||
U94353.1 | - | 4, 5 | 501 |