Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013829.2 | NP_001013851.2 | 5 | 891 | ||
GenBank mRNA | AK217249.1 | - | 1 | 348 | ||
BC042839.1 | - | 5 | 779 | 4488005 | ||
CB056323.1 | - | 4 | 505 |