Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002927.4 | NP_002918.1 | 6 | 465 | ||
NM_144766.2 | NP_658912.1 | 5 | 425 | |||
GenBank mRNA | AF030107.1 | - | 6 | 473 | ||
AF493935.1 | - | 3 | 170 | |||
AY562947.1 | - | 3 | 170 | |||
BC016667.1 | - | 5 | 391 | 4340668 | ||
BC056866.1 | - | 6 | 464 | 4419604 | ||
BT006929.1 | - | 3 | 170 | |||
BU942880.1 | - | 5 | 428 | |||
CR536532.1 | - | 3 | 170 |