Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007552.1 | NP_001007553.1 | 6 | 667 | ||
GenBank mRNA | AY217760.1 | - | 6 | 667 |
Gene Symbol: Itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010584.3 | NP_034714.1 | 6 | 677 | ||
GenBank mRNA | AB016496.1 | - | 6 | 676 | ||
AK007447.1 | - | 6 | 677 | |||
AY157363.1 | - | 6 | 666 | |||
AY157364.1 | - | 6 | 661 | |||
AY619693.1 | - | 6 | 660 | |||
BC145377.1 | - | 6 | 623 | 9053935 | ||
BC150760.1 | - | 6 | 623 | 9087671 | ||
BC150795.1 | - | 6 | 619 | 9087706 |