Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008429.1 | NP_001008429.1 | 1 | 151 | ||
GenBank mRNA | BC139171.1 | - | 1 | 193 | 8862193 | |
BC139172.1 | - | 1 | 193 | 8862194 |
Set Membership: |
G-protein coupled receptor |