Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013360.2 | NP_001013378.2 | 8 | 1508 | ||
NM_001013362.2 | NP_001013380.2 | 9 | 1799 |
Gene Symbol: Nptxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030689.4 | NP_109614.2 | 5 | 1875 | ||
GenBank mRNA | AF316612.1 | - | 5 | 1857 | ||
AK006611.1 | - | 3 | 836 | |||
AK147546.1 | - | 5 | 1932 | |||
AK147605.1 | - | 5 | 1883 | |||
BC058962.1 | - | 5 | 1874 | 6826075 | ||
BC098199.1 | - | 5 | 1874 | 5705707 | ||
BC141096.1 | - | 5 | 1854 | 9055708 | ||
BM944729.1 | - | 1 | 36 |