Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034896.2 | NP_001030068.1 | 3, 4 | 457 | ||
NM_001163172.1 | NP_001156644.1 | 3, 4 | 457 | |||
GenBank mRNA | AK139848.1 | - | 3, 4 | 457 | ||
BC147511.1 | - | 3, 4 | 254 | |||
BC172064.1 | - | 3, 4 | 254 | 9054350 |