Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005615.4 | NP_005606.1 | 2 | 456 | ||
XM_017021566.1 | XP_016877055.1 | 3 | 343 | |||
XM_017021567.1 | XP_016877056.1 | 3 | 356 | |||
GenBank mRNA | AK313580.1 | - | 2 | 439 | ||
BC020848.1 | - | 2 | 456 | 4732021 | ||
CR456965.1 | - | 1 | 163 |