Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199199.1 | NP_001186128.1 | 4 | 454 | ||
NM_001319139.1 | NP_001306068.1 | 5 | 572 | |||
NM_001319140.1 | NP_001306069.1 | 5 | 572 | |||
NM_001319141.1 | NP_001306070.1 | 4 | 454 | |||
NM_001319142.1 | NP_001306071.1 | 3 | 328 | |||
NM_001625.3 | NP_001616.1 | 4 | 454 | |||
NM_013411.4 | NP_037543.1 | 4 | 454 | |||
GenBank mRNA | AB005621.1 | - | 4 | 409 | ||
AB005622.1 | - | 4 | 413 | |||
AB451267.1 | - | 4 | 371 | |||
AB451394.1 | - | 4 | 371 | |||
AK291676.1 | - | 4 | 391 | |||
AK295105.1 | - | 5 | 505 | |||
AK296863.1 | - | 4 | 388 | |||
AK307731.1 | - | 3 | 257 | |||
AL557311.3 | - | 4 | 428 | |||
AY080899.1 | - | 4 | 406 | |||
AY080900.1 | - | 5 | 492 | |||
BC009405.2 | - | 4 | 382 | 4304794 | ||
BC026705.1 | - | 4 | 390 | |||
BC070127.1 | - | 4 | 396 | 6663039 | ||
BC090040.1 | - | 4 | 396 | 6471696 | ||
BQ889289.1 | - | 5 | 538 | |||
BX504559.1 | - | 4 | 435 | |||
DC422465.1 | - | 4 | 454 | |||
HY061267.1 | - | 4 | 455 | |||
U39945.1 | - | 4 | 404 | |||
U54645.1 | - | 4 | 374 | |||
U84371.1 | - | 4 | 402 |