Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008208.4 | NP_032234.3 | 2 | 232 | ||
GenBank mRNA | AK033602.1 | - | 2 | 233 | ||
BC125378.1 | - | 2 | 230 | 40129893 | ||
BC125380.1 | - | 2 | 230 | 40129895 | ||
D86082.1 | - | 2 | 209 |
Gene Symbol: H2-T3-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY989873.1 | - | 1 | 382 |