Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000329.2 | NP_000320.1 | 6 | 618 | ||
XM_017002027.1 | XP_016857516.1 | 5 | 830 | |||
GenBank mRNA | AK289925.1 | - | 6 | 609 | ||
BC075035.2 | - | 6 | 610 | 30915627 | ||
BC075036.2 | - | 6 | 613 | 30915421 | ||
U18991.1 | - | 6 | 618 |