Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 2, 3 | 411 | ||
NM_001035006.2 | NP_001030178.1 | 2, 3 | 162 | |||
NM_001199340.1 | NP_001186269.1 | 3, 4 | 260 | |||
NM_001199341.1 | NP_001186270.1 | 2, 3 | 261 | |||
NM_001199342.1 | NP_001186271.1 | 3, 4 | 359 | |||
NM_001199343.1 | NP_001186272.1 | 3, 4 | 342 | |||
NM_001199344.1 | NP_001186273.1 | 2, 3 | 244 | |||
GenBank mRNA | AK311828.1 | - | 2, 3 | 76 | ||
BC000502.2 | - | 1, 2 | 314 | 2821464 | ||
BC017831.1 | - | 1 | 80 | 4251433 | ||
BC066323.1 | - | 1 | 76 | 4797583 | ||
BC066324.1 | - | 1 | 80 | 4799029 | ||
BC106031.1 | - | 1 | 47 | 4540700 | ||
BF670086.1 | - | 258 | ||||
BG497914.1 | - | 2, 3 | 327 | |||
BG758380.1 | - | 3, 4 | 278 | |||
BG827384.1 | - | 2, 3 | 162 | |||
BI086293.1 | - | 2, 3 | 133 | |||
BQ877039.1 | - | 3, 4 | 300 | |||
BQ882430.1 | - | 2, 3 | 227 | |||
CB178432.1 | - | 1, 2 | 327 | |||
CD386294.1 | - | 2, 3 | 181 | |||
CD387752.1 | - | 2, 3 | 266 | |||
X53777.1 | - | 1, 2 | 179 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 1, 2 | 54 | ||
GenBank mRNA | BX393840.2 | - | 1, 2 | 56 |