Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dok6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039173.1 | NP_001034262.1 | 3 | 278 | ||
GenBank mRNA | AB221044.1 | - | 3 | 278 | ||
AK140422.1 | - | 1 | 56 | |||
BC138955.1 | - | 3 | 329 | 8861977 | ||
BC138957.1 | - | 3 | 329 | 8861979 |