Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011561.2 | NP_035691.2 | 7 | 844 | ||
NM_172552.3 | NP_766140.2 | 7 | 943 | |||
GenBank mRNA | AF102875.1 | - | 5 | 516 | ||
AK053870.1 | - | 7 | 871 | |||
AK164090.1 | - | 7 | 804 | |||
AK164643.1 | - | 7 | 911 | |||
AK169768.1 | - | 7 | 867 | |||
BC010315.1 | - | 7 | 833 | 3595866 | ||
BC085470.1 | - | 7 | 890 | 6416434 | ||
BC085471.1 | - | 7 | 914 | 6816716 |
Gene Symbol: Gm9855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF069519.1 | - | 1 | 801 | ||
AK089915.1 | - | 1 | 844 |