Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027978.2 | NP_082254.2 | 5 | 757 | ||
GenBank mRNA | AK009092.1 | - | 5 | 759 | ||
BC050978.1 | - | 1 | 103 | |||
BC080773.1 | - | 5 | 752 | 30139077 | ||
CK626929.1 | - | 1 | 29 |