Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hunk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015755.2 | NP_056570.1 | 11 | 1650 | ||
GenBank mRNA | AF055919.2 | - | 11 | 1650 | ||
AF167987.1 | - | 11 | 1662 | |||
AK144639.1 | - | 11 | 1633 | |||
AK147425.1 | - | 11 | 2036 | |||
AK147734.1 | - | 11 | 1404 | |||
BC137788.1 | - | 11 | 1711 | 8860807 | ||
BC137790.1 | - | 11 | 1710 | 8860809 |