Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023.3 | NP_001014.1 | 2 | 276 | ||
NM_001146227.1 | NP_001139699.1 | 2 | 276 | |||
GenBank mRNA | AK130961.1 | - | 2 | 201 | ||
AK301342.1 | - | 2 | 205 | |||
AK311808.1 | - | 2 | 201 | |||
AV716603.1 | - | 205 | ||||
BC007507.2 | - | 2 | 185 | 3029762 | ||
BC087850.1 | - | 2 | 171 | 30386230 | ||
BQ064297.1 | - | 2 | 186 | |||
CB138358.1 | - | 2 | 276 | |||
L06498.1 | - | 2 | 191 |