Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159483.1 | NP_001152955.1 | 5 | 420 | ||
NM_009078.2 | NP_033104.2 | 5 | 434 | |||
GenBank mRNA | AK010440.1 | - | 5 | 401 | ||
AK075817.1 | - | 5 | 402 | |||
AK168232.1 | - | 4 | 359 | |||
AK168481.1 | - | 5 | 405 | |||
AV467392.1 | - | 2 | 62 | |||
BC010710.1 | - | 4 | 379 | 2648593 | ||
BC083131.1 | - | 4 | 381 | 6442192 | ||
BC087961.1 | - | 5 | 400 | 6761831 | ||
BC089549.1 | - | 5 | 401 | 6703351 | ||
BC092539.1 | - | 5 | 390 | 6594084 | ||
BU524941.1 | - | 5 | 436 | |||
CB203522.1 | - | 5 | 420 | |||
M62952.1 | - | 4 | 372 |