Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163288.2 | NP_001156760.2 | 11 | 1818 | ||
GenBank mRNA | AK041075.1 | - | 2 | 96 | ||
BC157977.1 | - | 11 | 1743 | 9088138 | ||
BC158124.1 | - | 11 | 1795 | 9007393 |