Gene Symbol: Ndufc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024220.2 | NP_077182.1 | 2, 3 | 509 | ||
GenBank mRNA | AK002373.1 | - | 2, 3 | 417 | ||
AK003380.2 | - | 2, 3 | 411 | |||
AK008502.1 | - | 2, 3 | 424 | |||
AK008889.1 | - | 2, 3 | 417 | |||
AK019002.1 | - | 2, 3 | 436 | |||
AK153547.1 | - | 2, 3 | 439 | |||
AK166594.1 | - | 2, 3 | 436 | |||
AK168342.1 | - | 2, 3 | 461 | |||
BC002097.1 | - | 2, 3 | 432 | 3497384 | ||
CK128102.1 | - | 2, 3 | 509 |