Gene Symbol: Cypt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039942.2 | NP_001035031.1 | 1 | 214 | ||
GenBank mRNA | AM040458.1 | - | 1 | 214 |
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173412.2 | NP_775588.2 | 1 | 230 | ||
GenBank mRNA | AF463501.1 | - | 1 | 230 | ||
AK005791.1 | - | 1 | 222 | |||
AM040453.1 | - | 1 | 214 | |||
BC048530.1 | - | 1 | 219 | |||
BC060952.1 | - | 1 | 219 | 6772242 | ||
BC147783.1 | - | 1 | 314 | 9087502 |