Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017369.2 | NP_001017369.1 | 4 | 500 | ||
NM_006745.4 | NP_006736.1 | 5 | 786 | |||
XM_005263176.2 | XP_005263233.1 | 5 | 903 | |||
GenBank mRNA | AK292418.1 | - | 5 | 731 | ||
AK295432.1 | - | 4 | 446 | |||
AK309206.1 | - | 5 | 731 | |||
BC010653.1 | - | 4 | 680 | 3866053 | ||
BC107879.1 | - | 5 | 748 | 6498540 | ||
BU940384.1 | - | 461 | ||||
BX441001.2 | - | 5 | 786 | |||
U60205.1 | - | 4 | 682 | |||
U93162.1 | - | 5 | 746 |