Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002974.3 | NP_002965.1 | 7, 8 | 907 | ||
NM_175041.1 | NP_778206.1 | 7, 8 | 844 | |||
XM_011526138.1 | XP_011524440.1 | 7, 8 | 917 | |||
GenBank mRNA | AB046400.1 | - | 6, 7 | 712 | ||
AF461147.1 | - | 6, 7 | 770 | |||
AK292212.1 | - | 7, 8 | 907 | |||
AY245780.1 | - | 6, 7 | 770 | |||
AY245782.1 | - | 6, 7 | 770 | |||
BC017401.1 | - | 7, 8 | 847 | 4047303 | ||
U19557.1 | - | 7, 8 | 820 | |||
X89015.1 | - | 6, 7 | 770 |
Gene Symbol: SERPINB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF428138.1 | - | 7, 8 | 770 | ||
AF428139.1 | - | 7, 8 | 770 |