Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 2 | 755 | ||
GenBank mRNA | AB032261.1 | - | 2 | 645 | ||
AB208982.1 | - | 2 | 512 | |||
AF097514.1 | - | 2 | 500 | |||
AK222862.1 | - | 2 | 472 | |||
AK312312.1 | - | 2 | 537 | |||
BC005807.2 | - | 2 | 442 | |||
BC062303.1 | - | 2 | 641 | 6586153 | ||
BM546583.1 | - | 693 | ||||
S70284.1 | - | 1 | 260 | |||
Y13647.1 | - | 2 | 473 |