Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000695.3 | NP_000686.2 | 10 | 1514 | ||
NM_001031615.1 | NP_001026786.1 | 10 | 1358 | |||
GenBank mRNA | BC007685.2 | - | 10 | 1358 | 3636326 | |
BT006810.1 | - | 8 | 1075 |