Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 838 | ||
GenBank mRNA | AB232606.1 | - | 5 | 614 | ||
AK134125.1 | - | 5 | 583 | |||
AK146452.1 | - | 5 | 699 | |||
AK146460.1 | - | 5 | 650 | |||
AK146623.1 | - | 5 | 650 | |||
AK146978.1 | - | 5 | 630 | |||
AK147077.1 | - | 5 | 669 | |||
AK148022.1 | - | 5 | 711 | |||
AK148106.1 | - | 5 | 637 | |||
BC054753.1 | - | 5 | 654 | 6411651 | ||
BC085270.1 | - | 5 | 647 | 6478784 | ||
BC157916.1 | - | 5 | 667 | 9007344 | ||
L26528.1 | - | 5 | 649 |