Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 6, 7 | 1074 | ||
NM_201245.3 | NP_957697.1 | 6, 7 | 1074 | |||
GenBank mRNA | AK044512.1 | - | 6, 7 | 824 | ||
AK154807.1 | - | 6, 7 | 1081 | |||
AK155630.1 | - | 6, 7 | 855 | |||
AK158296.1 | - | 6, 7 | 1063 | |||
AK162530.1 | - | 3, 4 | 405 | |||
AK170849.1 | - | 6, 7 | 1058 | |||
AK171089.1 | - | 6, 7 | 1063 | |||
BC039932.1 | - | 5, 6 | 598 | |||
BC049803.1 | - | 6, 7 | 600 | |||
BC063067.1 | - | 6, 7 | 1037 | 5721396 | ||
U73200.1 | - | 6, 7 | 934 |