Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 4 | 721 | ||
NM_201245.3 | NP_957697.1 | 4 | 721 | |||
GenBank mRNA | AK044512.1 | - | 4 | 471 | ||
AK154807.1 | - | 4 | 728 | |||
AK155630.1 | - | 4 | 502 | |||
AK158296.1 | - | 4 | 710 | |||
AK162530.1 | - | 1 | 52 | |||
AK170849.1 | - | 4 | 705 | |||
AK171089.1 | - | 4 | 710 | |||
BC039932.1 | - | 3 | 245 | |||
BC049803.1 | - | 4 | 361 | |||
BC063067.1 | - | 4 | 684 | 5721396 | ||
U73200.1 | - | 4 | 581 |