Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004589.3 | NP_004580.1 | 2 | 401 | ||
XM_005256751.3 | XP_005256808.1 | 2 | 368 | |||
GenBank mRNA | AF026852.1 | - | 2 | 353 | ||
AF183424.1 | - | 2 | 353 | |||
AK074588.1 | - | 2 | 401 | |||
AK098826.1 | - | 2 | 361 | |||
AK315595.1 | - | 2 | 379 | |||
BC015504.2 | - | 2 | 356 | 3914068 | ||
DA571370.1 | - | 2 | 401 |