Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 2 | 209 | ||
NR_111964.1 | 2 | 401 | ||||
GenBank mRNA | AV739733.1 | - | 2 | 253 | ||
BC027888.1 | - | 2 | 197 | 5222247 | ||
BC107710.1 | - | 2 | 213 | 6699133 | ||
BC146914.1 | - | 2 | 160 | 9056842 | ||
X52149.1 | - | 2 | 387 |
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 2 | 199 | ||
GenBank mRNA | AI302038.1 | - | 2 | 100 | ||
BC146919.1 | - | 2 | 160 | 9056847 | ||
BC147008.1 | - | 2 | 160 | 9056936 | ||
BC147013.1 | - | 2 | 160 | 9056941 | ||
CF128666.1 | - | 2 | 178 |