Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 4 | 343 | ||
GenBank mRNA | AF088903.1 | - | 4 | 301 | ||
AF160355.1 | - | 4 | 289 | |||
AK080610.1 | - | 4 | 292 | |||
AK166069.1 | - | 4 | 293 | |||
AK167088.1 | - | 1 | 17 | |||
AK172879.1 | - | 3 | 206 | |||
AK192632.1 | - | 288 | ||||
BC014800.1 | - | 4 | 274 | 4220980 | ||
BY092839.1 | - | 4 | 318 | |||
CJ048539.1 | - | 4 | 344 |