Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 5 | 677 | ||
GenBank mRNA | AF088903.1 | - | 5 | 635 | ||
AF160355.1 | - | 5 | 623 | |||
AK077625.1 | - | 2 | 342 | |||
AK080610.1 | - | 5 | 626 | |||
AK166069.1 | - | 5 | 628 | |||
AK167088.1 | - | 2 | 351 | |||
AK172879.1 | - | 4 | 540 | |||
AK177992.1 | - | 2 | 200 | |||
AK196239.1 | - | 2 | 200 | |||
BC014800.1 | - | 5 | 608 | 4220980 |