Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twistnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172253.2 | NP_758457.1 | 3 | 560 | ||
GenBank mRNA | AK008677.1 | - | 3 | 542 | ||
AK010829.1 | - | 3 | 561 | |||
AK016638.1 | - | 3 | 557 | |||
AK017016.1 | - | 3 | 550 | |||
AK017810.1 | - | 3 | 545 | |||
AK153090.1 | - | 550 | ||||
AK160682.1 | - | 3 | 548 | |||
BC082580.1 | - | 532 | ||||
BC139162.1 | - | 3 | 548 | 8862184 | ||
BC139163.1 | - | 3 | 548 | 8862185 |