Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011482.4 | NP_035612.2 | 3 | 823 | ||
GenBank mRNA | AK004489.1 | - | 3 | 771 | ||
AK134943.1 | - | 761 | ||||
AK145391.1 | - | 3 | 825 | |||
AK145608.1 | - | 3 | 805 | |||
AK168808.1 | - | 3 | 793 | |||
AK168930.1 | - | 3 | 776 | |||
AK169103.1 | - | 3 | 807 | |||
BC026755.1 | - | 3 | 769 | 3154066 | ||
BC054450.1 | - | 3 | 769 | 6491298 | ||
BC083315.1 | - | 3 | 745 | 5714819 |