Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011482.4 | NP_035612.2 | 3 | 978 | ||
GenBank mRNA | AK004489.1 | - | 3 | 926 | ||
AK134943.1 | - | 916 | ||||
AK145391.1 | - | 3 | 980 | |||
AK145608.1 | - | 3 | 960 | |||
AK168930.1 | - | 3 | 931 | |||
AK169103.1 | - | 3 | 962 | |||
BC026755.1 | - | 3 | 924 | 3154066 | ||
BC054450.1 | - | 3 | 924 | 6491298 | ||
BC083315.1 | - | 3 | 900 | 5714819 |