Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011200.2 | NP_035330.1 | 3 | 829 | ||
GenBank mRNA | AK081491.1 | - | 2 | 295 | ||
AK148288.1 | - | 3 | 825 | |||
AK150506.1 | - | 2 | 459 | |||
AK150556.1 | - | 1 | 1412 | |||
AK150762.1 | - | 1 | 1413 | |||
AK151003.1 | - | 1 | 1413 | |||
AK151533.1 | - | 2 | 459 | |||
AK198788.1 | - | 2 | 110 | |||
BC003761.1 | - | 3 | 706 | |||
BC031734.1 | - | 3 | 751 | |||
BC055039.1 | - | 3 | 785 | 6396041 | ||
BC086787.1 | - | 2 | 135 | 30538771 | ||
BC094447.1 | - | 3 | 696 | 3990529 | ||
BC150972.1 | - | 2 | 501 | 9087885 | ||
BC150981.1 | - | 2 | 504 | 9087894 | ||
U84411.1 | - | 3 | 838 |