Search Thermo Fisher Scientific
Gene Symbol: CCL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002991.2 | NP_002982.2 | 2 | 139 | ||
XM_011516459.2 | XP_011514761.1 | 3 | 233 | |||
XM_011516460.2 | XP_011514762.1 | 4 | 406 | |||
GenBank mRNA | AJ223461.1 | - | 2 | 133 | ||
AK312216.1 | - | 3 | 233 | |||
BC069072.1 | - | 2 | 139 | 7216886 | ||
BC069391.1 | - | 2 | 139 | 7262740 | ||
U85768.1 | - | 2 | 139 |