Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163575.1 | NP_001157047.1 | 8 | 1990 | ||
NM_001163576.1 | NP_001157048.1 | 7 | 2094 | |||
GenBank mRNA | AK028373.1 | - | 8 | 1938 | ||
AK035309.1 | - | 7 | 2114 | |||
AK046755.1 | - | 7 | 2075 | |||
AK146336.1 | - | 8 | 1990 | |||
AK153205.1 | - | 7 | 1831 | |||
AK166291.1 | - | 8 | 1941 | |||
BC023056.1 | - | 4 | 827 | |||
BC024074.1 | - | 8 | 1947 | |||
BC025608.1 | - | 4 | 972 |