Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194336.2 | NP_919317.2 | 11 | 1836 | ||
GenBank mRNA | AK128993.1 | - | 1832 | |||
AK129021.1 | - | 1108 | ||||
AK171913.1 | - | 8 | 3474 | |||
BC007143.1 | - | 10 | 1693 | 3495728 | ||
BC052867.1 | - | 6 | 1064 | |||
BC057969.1 | - | 1822 | 4978977 | |||
BC115768.1 | - | 1832 | 40085916 |
Gene Symbol: Gbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039646.2 | NP_001034735.2 | 11 | 1842 | ||
GenBank mRNA | BC112328.1 | - | 11 | 1859 | 5348193 | |
BC114209.1 | - | 11 | 1842 | 4980612 | ||
DQ985743.1 | - | 11 | 1842 |