Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005065.5 | NP_005056.3 | 17 | 1798 | ||
XM_005267988.2 | XP_005268045.1 | 17 | 1681 | |||
XM_005267989.3 | XP_005268046.1 | 16 | 1732 | |||
GenBank mRNA | AB020335.1 | - | 17 | 1726 | ||
AF052059.1 | - | 17 | 1774 | |||
AK075511.1 | - | 17 | 1777 |