Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005065.5 | NP_005056.3 | 19 | 2050 | ||
XM_005267988.2 | XP_005268045.1 | 19 | 1933 | |||
XM_005267989.3 | XP_005268046.1 | 18 | 1984 | |||
GenBank mRNA | AB020335.1 | - | 19 | 1978 | ||
AF052059.1 | - | 19 | 2026 | |||
BC040498.1 | - | 2 | 137 |