Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011792.1 | NP_001011792.1 | 1 | 803 |
Gene Symbol: Olfr208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146384.1 | NP_666496.1 | 1 | 803 |