Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adra2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138506.1 | NP_612515.1 | 1 | 1378 | ||
GenBank mRNA | M58316.1 | - | 1 | 1378 |
Set Membership: |
![]() |