Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031684.2 | NP_001026854.1 | 2 | 408 | ||
NM_001195446.1 | NP_001182375.1 | 2 | 408 | |||
XM_005264484.2 | XP_005264541.1 | 2 | 275 | |||
XM_005264485.2 | XP_005264542.1 | 2 | 275 | |||
GenBank mRNA | AB445102.1 | - | 2 | 170 | ||
AK091425.1 | - | 2 | 223 | |||
AK225141.1 | - | 2 | 275 | |||
AK293667.1 | - | 2 | 275 | |||
AY166860.1 | - | 2 | 296 | |||
AY513287.1 | - | 2 | 251 | |||
BC000997.2 | - | 2 | 255 | 2901284 | ||
BC017369.2 | - | 2 | 266 | 2967417 | ||
BC017908.1 | - | 2 | 273 | 4276642 | ||
BC022328.1 | - | 2 | 273 | 4251314 | ||
BT006745.1 | - | 2 | 170 | |||
DC358626.1 | - | 2 | 409 | |||
L22253.1 | - | 2 | 275 |