Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012499.1 | NP_036631.1 | 9 | 1291 | ||
XM_006254506.2 | XP_006254568.1 | 9 | 1497 | |||
XM_006254508.2 | XP_006254570.1 | 10 | 1338 | |||
XM_006254509.2 | XP_006254571.1 | 10 | 1307 | |||
XM_006254510.2 | XP_006254572.1 | 9 | 1497 | |||
XM_006254511.2 | XP_006254573.1 | 10 | 1338 | |||
XM_006254512.2 | XP_006254574.1 | 9 | 1946 | |||
XM_006254513.2 | XP_006254575.1 | 8 | 1392 | |||
XM_006254515.2 | XP_006254577.1 | 10 | 1163 | |||
XM_006254518.2 | XP_006254580.1 | 9 | 1761 | |||
XM_008772015.1 | XP_008770237.1 | 9 | 3429 | |||
XM_008772016.1 | XP_008770238.1 | 9 | 3379 | |||
XM_008772017.1 | XP_008770239.1 | 9 | 1770 | |||
GenBank mRNA | D38629.1 | - | 9 | 1291 |