Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271053.1 | NP_001257982.1 | 3 | 286 | ||
NM_012501.2 | NP_036633.2 | 3 | 243 | |||
XM_008766144.1 | XP_008764366.1 | 3 | 278 | |||
XM_008766146.1 | XP_008764368.1 | 3 | 256 | |||
GenBank mRNA | AW917416.1 | - | 3 | 195 | ||
FQ209522.1 | - | 3 | 210 | |||
FQ209891.1 | - | 3 | 212 | |||
FQ210075.1 | - | 3 | 243 | |||
FQ210160.1 | - | 3 | 212 | |||
FQ210397.1 | - | 3 | 210 | |||
FQ210428.1 | - | 3 | 214 | |||
FQ210554.1 | - | 3 | 211 | |||
FQ210884.1 | - | 3 | 209 | |||
FQ211165.1 | - | 3 | 210 | |||
FQ211318.1 | - | 3 | 212 | |||
FQ211343.1 | - | 3 | 212 | |||
FQ211392.1 | - | 3 | 214 | |||
FQ211416.1 | - | 3 | 215 | |||
FQ211454.1 | - | 3 | 214 | |||
FQ218088.1 | - | 3 | 242 | |||
FQ218207.1 | - | 3 | 242 | |||
FQ218211.1 | - | 3 | 286 | |||
FQ218286.1 | - | 3 | 213 | |||
FQ219230.1 | - | 3 | 210 |