Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270630.1 | NP_001257559.1 | 2 | 738 | ||
NM_001270631.1 | NP_001257560.1 | 2 | 889 | |||
NM_001270632.1 | NP_001257561.1 | 2 | 806 | |||
NM_001270633.1 | NP_001257562.1 | 2 | 730 | |||
NM_001270634.1 | NP_001257563.1 | 3 | 617 | |||
NM_001270635.1 | NP_001257564.1 | 2 | 616 | |||
NM_001270636.1 | NP_001257565.1 | 2 | 602 | |||
NM_001270637.1 | NP_001257566.1 | 2 | 594 | |||
NM_001270638.1 | NP_001257567.1 | 2 | 471 | |||
NM_012513.4 | NP_036645.2 | 2 | 1029 | |||
XM_006234684.2 | XP_006234746.1 | 2 | 737 | |||
XM_008762078.1 | XP_008760300.1 | 2 | 2663 | |||
GenBank mRNA | AY176065.1 | - | 1 | 369 | ||
AY559248.1 | - | 2 | 532 | |||
AY559249.1 | - | 2 | 546 | |||
AY559250.1 | - | 3 | 618 | |||
BC087634.1 | - | 2 | 724 | 7319966 | ||
EF125675.1 | - | 2 | 1029 | |||
EF125676.1 | - | 2 | 595 | |||
EF125677.1 | - | 2 | 807 | |||
EF125678.1 | - | 2 | 890 | |||
EF125679.1 | - | 2 | 730 | |||
EF125680.1 | - | 2 | 738 | |||
EF125686.1 | - | 2 | 616 | |||
EF125687.1 | - | 2 | 471 | |||
EF125688.1 | - | 2 | 603 | |||
EF125689.1 | - | 2 | 667 | |||
EF125690.1 | - | 1 | 861 | |||
M61175.1 | - | 2 | 441 | |||
X67108.1 | - | 1 | 390 |