Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012519.2 | NP_036651.1 | 16, 17 | 1368 | ||
XM_006233284.2 | XP_006233346.1 | 15, 16 | 1527 | |||
XM_006233285.2 | XP_006233347.1 | 15, 16 | 1518 | |||
XM_008761452.1 | XP_008759674.1 | 16, 17 | 1587 | |||
GenBank mRNA | BC107562.1 | - | 1128 | 7939424 | ||
J05072.1 | - | 1368 | ||||
L13406.1 | - | 2, 3 | 105 | |||
L13407.1 | - | 3, 4 | 138 | |||
L13408.1 | - | 3, 4 | 165 |