Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301715.1 | NP_001288644.1 | 1 | 1391 | ||
NM_001301720.1 | NP_001288649.1 | 1 | 1391 | |||
NM_024125.5 | NP_077039.3 | 1 | 1391 | |||
GenBank mRNA | BC129071.1 | - | 1 | 1381 | 7458221 | |
M57235.1 | - | 1 | 1378 | |||
X60769.1 | - | 1 | 1110 |